Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGCTGCCCCACTTCGACCTGAGC[G/T]GGGAGGCCACTAGCAGCCAGTTCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608919 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTYH3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TTYH3 - tweety family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025250.2 | 228 | Missense Mutation | GGG,TGG | G,W 25 | NP_079526.1 | |
XM_011515535.2 | 228 | Missense Mutation | GGG,TGG | G,W 25 | XP_011513837.1 | |
XM_017012655.1 | 228 | Missense Mutation | GGG,TGG | G,W 25 | XP_016868144.1 | |
XM_017012656.1 | 228 | Missense Mutation | GGG,TGG | G,W 25 | XP_016868145.1 |