Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACTGGTTGGAGGTCGGAGGGTGG[C/G]CATGAGGGTGGTGGCTGAGCGAGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600028 MIM: 600030 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DLX5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DLX5 - distal-less homeobox 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005221.5 | 2035 | Missense Mutation | GCC,GGC | A,G 239 | NP_005212.1 | |
XM_005250185.3 | 2035 | Missense Mutation | GCC,GGC | A,G 111 | XP_005250242.1 | |
XM_017011803.1 | 2035 | Missense Mutation | GCC,GGC | A,G 111 | XP_016867292.1 |
DLX6 - distal-less homeobox 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DLX6-AS1 - DLX6 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |