Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACGAGTGCCCGCGCACCGTGCGCA[A/G]CGGCGACTTCGTGCGCTACCACTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616257 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FKBP9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FKBP9 - FK506 binding protein 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284341.1 | 328 | Missense Mutation | AAC,AGC | N,S 53 | NP_001271270.1 | |
NM_001284343.1 | 328 | Intron | NP_001271272.1 | |||
NM_007270.4 | 328 | Missense Mutation | AAC,AGC | N,S 53 | NP_009201.2 | |
XM_011515115.2 | 328 | Missense Mutation | AAC,AGC | N,S 53 | XP_011513417.1 |