Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGCGTTACAGCCCAAAGATGAT[A/G]TTGTAGACCGAGCAAAAATGGAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600287 | ||||||||||||||||||||
Literature Links: |
GARS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GARS - glycyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316772.1 | 539 | Missense Mutation | ATT,GTT | I,V 64 | NP_001303701.1 | |
NM_002047.3 | 539 | Missense Mutation | ATT,GTT | I,V 118 | NP_002038.2 | |
XM_006715686.2 | 539 | UTR 5 | XP_006715749.1 |