Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGACTCTTGGCACATAAGTCTGG[A/T]GACTGCTGTGGGGGTGGCAGTGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605341 | ||||||||||||||||||||
Literature Links: |
PILRA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PILRA - paired immunoglobin like type 2 receptor alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013439.2 | 796 | Missense Mutation | GAG,GTG | E,V 195 | NP_038467.2 | |
NM_178272.1 | 796 | Intron | NP_840056.1 | |||
NM_178273.1 | 796 | Intron | NP_840057.1 |