Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAAAGCTGAGTAGTGAGGCTGCC[A/G]CTGGTCCAGGAGAGGGTGTGAGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600011 MIM: 602372 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPHB4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPHB4 - EPH receptor B4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004444.4 | 2852 | Missense Mutation | CGG,TGG | R,W 902 | NP_004435.3 | |
XM_017011816.1 | 2852 | Missense Mutation | CGG,TGG | R,W 920 | XP_016867305.1 |
ZAN - zonadhesin (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |