Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTCCTCCGAGGGGATCAGCATCC[A/G]CTTCTCCAGGCTCCGGCTCTTCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600197 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAFK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAFK - MAF bZIP transcription factor K | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM184A - transmembrane protein 184A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001097620.1 | 2522 | Silent Mutation | NP_001091089.1 | |||
XM_006715653.1 | 2522 | Missense Mutation | XP_006715716.1 | |||
XM_006715655.2 | 2522 | Silent Mutation | XP_006715718.1 | |||
XM_011515164.1 | 2522 | Missense Mutation | XP_011513466.1 | |||
XM_011515165.2 | 2522 | Intron | XP_011513467.1 | |||
XM_011515166.2 | 2522 | Intron | XP_011513468.1 | |||
XM_017011815.1 | 2522 | UTR 5 | XP_016867304.1 |