Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATTACATTCATATTGCCTCTCCT[A/G]TGAATTCTCTGATGGCTGTTGAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194536 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF12 - zinc finger protein 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006956.2 | 2489 | Silent Mutation | CAC,CAT | H,H 645 | NP_008887.2 | |
NM_016265.3 | 2489 | Silent Mutation | CAC,CAT | H,H 683 | NP_057349.2 |