Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCATCCTTGGAGACAGAAGCCAT[C/T]CTGCAACACAGAAAATCACAGCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605842 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TBL2 - transducin (beta)-like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012453.2 | 883 | Silent Mutation | AGA,AGG | R,R 293 | NP_036585.1 | |
XM_006715923.3 | 883 | Silent Mutation | AGA,AGG | R,R 260 | XP_006715986.1 | |
XM_011516027.2 | 883 | Silent Mutation | AGA,AGG | R,R 128 | XP_011514329.1 |