Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAATGTGGCCAAGAGTTACCTTC[C/G]TAATGCACACCTGGAATGTCATTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605326 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAX1BP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TAX1BP1 - Tax1 binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079864.2 | 265 | Missense Mutation | CCT,CGT | P,R 28 | NP_001073333.1 | |
NM_001206901.1 | 265 | Missense Mutation | CCT,CGT | P,R 28 | NP_001193830.1 | |
NM_001206902.1 | 265 | UTR 5 | NP_001193831.1 | |||
NM_006024.6 | 265 | Missense Mutation | CCT,CGT | P,R 28 | NP_006015.4 | |
XM_005249900.2 | 265 | Missense Mutation | CCT,CGT | P,R 28 | XP_005249957.1 | |
XM_017012754.1 | 265 | Missense Mutation | CCT,CGT | P,R 28 | XP_016868243.1 |