Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGAAATCGGTGCAGGTGGGAGA[C/G]GGCGGCCAGCATCAGTGCCCCCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609721 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C7orf69 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C7orf69 - chromosome 7 open reading frame 69 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302627.1 | 8024 | Intron | NP_001289556.1 | |||
NM_025031.2 | 8024 | Intron | NP_079307.2 |
PKD1L1 - polycystin 1 like 1, transient receptor potential channel interacting | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138295.3 | 8024 | Missense Mutation | CTC,GTC | L,V 2664 | NP_612152.1 | |
XM_017011798.1 | 8024 | Missense Mutation | CTC,GTC | L,V 2723 | XP_016867287.1 |