Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCGGTGCCGATCAGCGGAAGACC[A/G]AAGAGGATGAGAAGGACGACAAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605502 MIM: 154365 MIM: 604943 | ||||||||||||||||||||
Literature Links: |
DNAJC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAJC2 - DnaJ heat shock protein family (Hsp40) member C2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSMC2 - proteasome 26S subunit, ATPase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204453.1 | 346 | Missense Mutation | AAA,GAA | K,E 13 | NP_001191382.1 | |
NM_002803.3 | 346 | Missense Mutation | AAA,GAA | K,E 13 | NP_002794.1 | |
XM_005250505.1 | 346 | Missense Mutation | AAA,GAA | K,E 13 | XP_005250562.1 |
SLC26A5 - solute carrier family 26 member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |