Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCAGGGGATTATGGGCACCTACC[A/G]TCCCTGGGCCGAGGAGTGGGCACCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607639 MIM: 182889 | ||||||||||||||||||||
Literature Links: |
SSC4D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SSC4D - scavenger receptor cysteine rich family member with 4 domains | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080744.1 | 1547 | Silent Mutation | GAC,GAT | D,D 470 | NP_542782.1 | |
XM_017011750.1 | 1547 | Silent Mutation | GAC,GAT | D,D 301 | XP_016867239.1 |
ZP3 - zona pellucida glycoprotein 3 (sperm receptor) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |