Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGATAATCAGCTCAACACTGTC[C/G]TCACCATCAAGTAACGCCCCAGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615753 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
POM121 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
POM121 - POM121 transmembrane nucleoporin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257190.2 | Intron | NP_001244119.1 | ||||
NM_172020.4 | Intron | NP_742017.1 | ||||
XM_005250726.3 | Intron | XP_005250783.1 | ||||
XM_005250727.3 | Intron | XP_005250784.1 | ||||
XM_006716196.3 | Intron | XP_006716259.1 | ||||
XM_011516730.1 | Intron | XP_011515032.1 | ||||
XM_017012853.1 | Intron | XP_016868342.1 | ||||
XM_017012854.1 | Intron | XP_016868343.1 |