Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCATTCTCCAGAATTCAATTTGTT[C/T]TCCAACTCGGTGGTGTTTGAAAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM71F2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM71F2 - family with sequence similarity 71 member F2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012454.5 | 249 | Silent Mutation | TTC,TTT | F,F 39 | NP_001012457.3 | |
NM_001128926.3 | 249 | Silent Mutation | TTC,TTT | F,F 39 | NP_001122398.1 | |
NM_001290254.1 | 249 | UTR 5 | NP_001277183.1 | |||
NM_001290255.1 | 249 | UTR 5 | NP_001277184.1 | |||
XM_011516128.2 | 249 | Silent Mutation | TTC,TTT | F,F 39 | XP_011514430.1 | |
XM_011516129.2 | 249 | Silent Mutation | TTC,TTT | F,F 39 | XP_011514431.1 | |
XM_011516130.2 | 249 | Silent Mutation | TTC,TTT | F,F 39 | XP_011514432.1 | |
XM_011516131.2 | 249 | Silent Mutation | TTC,TTT | F,F 39 | XP_011514433.1 | |
XM_011516132.2 | 249 | Silent Mutation | TTC,TTT | F,F 39 | XP_011514434.1 | |
XM_011516134.2 | 249 | UTR 5 | XP_011514436.1 | |||
XM_011516135.2 | 249 | Intron | XP_011514437.1 | |||
XM_011516136.2 | 249 | Intron | XP_011514438.1 | |||
XM_017012147.1 | 249 | UTR 5 | XP_016867636.1 | |||
XM_017012149.1 | 249 | Silent Mutation | TTC,TTT | F,F 39 | XP_016867638.1 |