Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAGGTGATGGATGTTTTCCTCC[A/G]GTTCTCAGGGCTCCACCTCTTTCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608037 MIM: 615245 MIM: 601737 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHPF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHPF2 - chondroitin polymerizing factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284295.1 | 3652 | Missense Mutation | CAG,CGG | Q,R 702 | NP_001271224.1 | |
NM_019015.2 | 3652 | Missense Mutation | CAG,CGG | Q,R 710 | NP_061888.1 |
MIR671 - microRNA 671 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMARCD3 - SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003801.1 | 3652 | Intron | NP_001003801.1 | |||
NM_001003802.1 | 3652 | Intron | NP_001003802.1 | |||
NM_003078.3 | 3652 | Intron | NP_003069.2 | |||
XM_011516521.2 | 3652 | Intron | XP_011514823.1 | |||
XM_017012556.1 | 3652 | Intron | XP_016868045.1 |