Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGCACGTGTGGGTTCAGGATGGT[C/T]GTGTGGATCGGCTTGCTGTTCTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607707 MIM: 606209 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAMK2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAMK2B - calcium/calmodulin dependent protein kinase II beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001220.4 | 2015 | Silent Mutation | ACA,ACG | T,T 600 | NP_001211.3 | |
NM_001293170.1 | 2015 | Silent Mutation | ACA,ACG | T,T 476 | NP_001280099.1 | |
NM_172078.2 | 2015 | Silent Mutation | ACA,ACG | T,T 476 | NP_742075.1 | |
NM_172079.2 | 2015 | Silent Mutation | ACA,ACG | T,T 452 | NP_742076.1 | |
NM_172080.2 | 2015 | Silent Mutation | ACA,ACG | T,T 451 | NP_742077.1 | |
NM_172081.2 | 2015 | Silent Mutation | ACA,ACG | T,T 437 | NP_742078.1 | |
NM_172082.2 | 2015 | Silent Mutation | ACA,ACG | T,T 426 | NP_742079.1 | |
NM_172083.2 | 2015 | Silent Mutation | ACA,ACG | T,T 413 | NP_742080.1 | |
NM_172084.2 | 2015 | Silent Mutation | ACA,ACG | T,T 383 | NP_742081.1 | |
XM_005249862.3 | 2015 | Silent Mutation | ACA,ACG | T,T 461 | XP_005249919.1 | |
XM_005249864.3 | 2015 | Silent Mutation | ACA,ACG | T,T 428 | XP_005249921.1 | |
XM_006715781.2 | 2015 | Silent Mutation | ACA,ACG | T,T 560 | XP_006715844.1 | |
XM_006715784.2 | 2015 | Silent Mutation | ACA,ACG | T,T 452 | XP_006715847.1 | |
XM_011515547.1 | 2015 | Silent Mutation | ACA,ACG | T,T 643 | XP_011513849.1 | |
XM_011515549.1 | 2015 | Silent Mutation | ACA,ACG | T,T 628 | XP_011513851.1 | |
XM_011515550.1 | 2015 | Silent Mutation | ACA,ACG | T,T 619 | XP_011513852.1 | |
XM_011515551.1 | 2015 | Silent Mutation | ACA,ACG | T,T 619 | XP_011513853.1 | |
XM_011515552.1 | 2015 | Silent Mutation | ACA,ACG | T,T 618 | XP_011513854.1 | |
XM_011515553.1 | 2015 | Silent Mutation | ACA,ACG | T,T 605 | XP_011513855.1 | |
XM_011515554.1 | 2015 | Silent Mutation | ACA,ACG | T,T 603 | XP_011513856.1 | |
XM_011515555.1 | 2015 | Silent Mutation | ACA,ACG | T,T 600 | XP_011513857.1 | |
XM_011515556.1 | 2015 | Silent Mutation | ACA,ACG | T,T 595 | XP_011513858.1 | |
XM_011515557.1 | 2015 | Silent Mutation | ACA,ACG | T,T 594 | XP_011513859.1 | |
XM_011515558.2 | 2015 | Intron | XP_011513860.1 | |||
XM_011515559.1 | 2015 | Silent Mutation | ACA,ACG | T,T 514 | XP_011513861.1 | |
XM_017012660.1 | 2015 | Silent Mutation | ACA,ACG | T,T 604 | XP_016868149.1 | |
XM_017012661.1 | 2015 | Silent Mutation | ACA,ACG | T,T 590 | XP_016868150.1 | |
XM_017012662.1 | 2015 | Silent Mutation | ACA,ACG | T,T 537 | XP_016868151.1 | |
XM_017012663.1 | 2015 | Silent Mutation | ACA,ACG | T,T 490 | XP_016868152.1 | |
XM_017012664.1 | 2015 | Silent Mutation | ACA,ACG | T,T 436 | XP_016868153.1 | |
XM_017012665.1 | 2015 | Silent Mutation | ACA,ACG | T,T 427 | XP_016868154.1 | |
XM_017012666.1 | 2015 | Silent Mutation | ACA,ACG | T,T 347 | XP_016868155.1 | |
XM_017012667.1 | 2015 | Silent Mutation | ACA,ACG | T,T 332 | XP_016868156.1 | |
XM_017012668.1 | 2015 | Silent Mutation | ACA,ACG | T,T 323 | XP_016868157.1 | |
XM_017012669.1 | 2015 | Silent Mutation | ACA,ACG | T,T 323 | XP_016868158.1 | |
XM_017012670.1 | 2015 | Silent Mutation | ACA,ACG | T,T 308 | XP_016868159.1 |
YKT6 - YKT6 v-SNARE homolog (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |