Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCAATGCCTGGACACTGCGCAGC[A/G]GAATTTATATAGAGATGTGATGTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603974 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF92 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF92 - zinc finger protein 92 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287532.1 | 276 | Missense Mutation | CAG,CGG | Q,R 26 | NP_001274461.1 | |
NM_001287533.1 | 276 | UTR 5 | NP_001274462.1 | |||
NM_001287534.1 | 276 | Intron | NP_001274463.1 | |||
NM_007139.3 | 276 | Intron | NP_009070.2 | |||
NM_152626.3 | 276 | Missense Mutation | CAG,CGG | Q,R 26 | NP_689839.1 |