Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAGCCGCGAGCGCAGCGGAAAT[A/C]GGGCTCTGGCTGCTCCGGCCTGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600642 | ||||||||||||||||||||
Literature Links: |
MYO1G PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MYO1G - myosin IG | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033054.2 | 2910 | Missense Mutation | GAT,TAT | D,Y 1003 | NP_149043.2 | |
XM_017012503.1 | 2910 | Missense Mutation | GAT,TAT | D,Y 888 | XP_016867992.1 | |
XM_017012504.1 | 2910 | Missense Mutation | GAT,TAT | D,Y 765 | XP_016867993.1 |