Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCACGGGCTTCGTGCTGGTGCCC[C/T]TCTCTGGCTACCTGGTGCTGGGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605681 MIM: 601766 | ||||||||||||||||||||
Literature Links: |
BAZ1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BAZ1B - bromodomain adjacent to zinc finger domain 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FZD9 - frizzled class receptor 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003508.2 | 1445 | Missense Mutation | CTC,TTC | L,F 406 | NP_003499.1 |