Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGTCTCAGCAGAAGCTGCTTCTC[A/G]TGAATGAGCCAGAACCAGTCCACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MICALL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MICALL2 - MICAL like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182924.3 | 2529 | Silent Mutation | CAC,CAT | H,H 786 | NP_891554.1 | |
XM_011515520.2 | 2529 | Silent Mutation | CAC,CAT | H,H 786 | XP_011513822.1 | |
XM_011515522.2 | 2529 | Silent Mutation | CAC,CAT | H,H 786 | XP_011513824.1 | |
XM_011515523.2 | 2529 | Silent Mutation | CAC,CAT | H,H 703 | XP_011513825.1 | |
XM_011515524.2 | 2529 | Silent Mutation | CAC,CAT | H,H 215 | XP_011513826.1 | |
XM_017012619.1 | 2529 | Intron | XP_016868108.1 |