Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGCGTTGACTGCTACTATGTCTC[C/T]TCTCACAGATATGCCCATTTCTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176842 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C7orf25 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C7orf25 - chromosome 7 open reading frame 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099858.1 | 765 | Missense Mutation | AGA,GGA | R,G 248 | NP_001093328.1 | |
NM_024054.2 | 765 | Missense Mutation | AGA,GGA | R,G 190 | NP_076959.2 | |
XM_005249844.3 | 765 | Missense Mutation | AGA,GGA | R,G 190 | XP_005249901.1 | |
XM_017012596.1 | 765 | Missense Mutation | AGA,GGA | R,G 204 | XP_016868085.1 | |
XM_017012597.1 | 765 | Missense Mutation | AGA,GGA | R,G 190 | XP_016868086.1 |
PSMA2 - proteasome subunit alpha 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |