Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTGTACCTGATAAATCAAAACCA[C/T]CACCACAAAAGGATCAAGGTACTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
USP42 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
USP42 - ubiquitin specific peptidase 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032172.2 | 288 | Missense Mutation | CCA,TCA | P,S 75 | NP_115548.1 | |
XM_005249883.4 | 288 | Missense Mutation | CCA,TCA | P,S 75 | XP_005249940.1 | |
XM_006715790.1 | 288 | Missense Mutation | CCA,TCA | P,S 75 | XP_006715853.1 | |
XM_006715791.2 | 288 | Missense Mutation | CCA,TCA | P,S 75 | XP_006715854.1 | |
XM_011515573.1 | 288 | Missense Mutation | CCA,TCA | P,S 75 | XP_011513875.1 | |
XM_011515574.1 | 288 | UTR 5 | XP_011513876.1 | |||
XM_011515577.1 | 288 | Intron | XP_011513879.1 | |||
XM_011515578.1 | 288 | UTR 5 | XP_011513880.1 |