Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGATCCCGGAGCGAGGCCACAGGC[A/G]GCGAGGCAGCTTCCGCTCCAGCATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602757 MIM: 606680 | ||||||||||||||||||||
Literature Links: |
EPHB6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EPHB6 - EPH receptor B6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRPV6 - transient receptor potential cation channel subfamily V member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018646.5 | 2060 | Missense Mutation | CGC,TGC | R,C 651 | NP_061116.5 |