Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTATTACAACTGGGGGCGGTTCTC[A/G]AAGCCCCCAGAAGGTGAGGCGAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605719 MIM: 600404 | ||||||||||||||||||||
Literature Links: |
LAT2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LAT2 - linker for activation of T-cells family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014146.3 | 765 | Silent Mutation | TCA,TCG | S,S 125 | NP_054865.2 | |
NM_032463.2 | 765 | Silent Mutation | TCA,TCG | S,S 125 | NP_115852.1 | |
NM_032464.2 | 765 | Silent Mutation | TCA,TCG | S,S 125 | NP_115853.2 | |
XM_011516558.2 | 765 | Silent Mutation | TCA,TCG | S,S 125 | XP_011514860.1 |
RFC2 - replication factor C subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |