Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCACGCGCTGTGCATGGAGGCGCC[C/T]GAGAACGCCACGGCCGGCCCCGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605681 MIM: 601766 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAZ1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BAZ1B - bromodomain adjacent to zinc finger domain 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FZD9 - frizzled class receptor 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003508.2 | 697 | Silent Mutation | CCC,CCT | P,P 156 | NP_003499.1 |