Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGGACAGACAGGCTGCGGGCCCC[A/G]ATATCCTGGCAGCCGTGTTGGATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 146690 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IMPDH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IMPDH1 - inosine monophosphate dehydrogenase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000883.3 | 1719 | Silent Mutation | NP_000874.2 | |||
NM_001102605.1 | 1719 | Silent Mutation | NP_001096075.1 | |||
NM_001142573.1 | 1719 | Intron | NP_001136045.1 | |||
NM_001142574.1 | 1719 | Intron | NP_001136046.1 | |||
NM_001142575.1 | 1719 | Intron | NP_001136047.1 | |||
NM_001142576.1 | 1719 | Silent Mutation | NP_001136048.1 | |||
NM_001304521.1 | 1719 | Silent Mutation | NP_001291450.1 | |||
NM_183243.2 | 1719 | Intron | NP_899066.1 | |||
XM_017012168.1 | 1719 | Silent Mutation | XP_016867657.1 | |||
XM_017012169.1 | 1719 | Silent Mutation | XP_016867658.1 | |||
XM_017012170.1 | 1719 | Silent Mutation | XP_016867659.1 | |||
XM_017012171.1 | 1719 | Silent Mutation | XP_016867660.1 | |||
XM_017012172.1 | 1719 | Silent Mutation | XP_016867661.1 | |||
XM_017012173.1 | 1719 | Intron | XP_016867662.1 |