Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCGGTGGGCAGCATGGACAGTGT[C/T]GGGGGTGGCCCTGGCGGGGCCAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615477 MIM: 611914 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NYAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NYAP1 - neuronal tyrosine phosphorylated phosphoinositide-3-kinase adaptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173564.3 | 496 | Silent Mutation | GTC,GTT | V,V 94 | NP_775835.2 | |
XM_006715907.3 | 496 | Silent Mutation | GTC,GTT | V,V 94 | XP_006715970.1 | |
XM_017011868.1 | 496 | Silent Mutation | GTC,GTT | V,V 94 | XP_016867357.1 |
TSC22D4 - TSC22 domain family member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |