Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATAACCAATTTGCCAAATGCACTA[A/G]GGGATATGTAATACGAGGAAAGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM168 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM168 - transmembrane protein 168 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287497.1 | 2340 | Silent Mutation | CTA,TTA | L,L 650 | NP_001274426.1 | |
NM_022484.5 | 2340 | Silent Mutation | CTA,TTA | L,L 650 | NP_071929.3 | |
XM_017012523.1 | 2340 | Silent Mutation | CTA,TTA | L,L 650 | XP_016868012.1 | |
XM_017012524.1 | 2340 | Intron | XP_016868013.1 | |||
XM_017012525.1 | 2340 | Silent Mutation | CTA,TTA | L,L 266 | XP_016868014.1 |