Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAATTCTGCGCTTCTCAGTAACT[A/T]CGAGGTAAATTGGATTGTTACATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606121 | ||||||||||||||||||||
Literature Links: |
CRCP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CRCP - CGRP receptor component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040647.1 | 186 | Missense Mutation | TAC,TTC | Y,F 14 | NP_001035737.1 | |
NM_001040648.1 | 186 | Intron | NP_001035738.1 | |||
NM_001142414.1 | 186 | Missense Mutation | TAC,TTC | Y,F 40 | NP_001135886.1 | |
NM_014478.4 | 186 | Missense Mutation | TAC,TTC | Y,F 14 | NP_055293.1 |