Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCTTCGTCTTCTGTTGTAGAAA[C/T]GTCTGAGCCCATGTTGTCGTGAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600514 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927870 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101927870 - uncharacterized LOC101927870 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RELN - reelin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005045.3 | 10500 | Missense Mutation | CAT,CGT | H,R 3447 | NP_005036.2 | |
NM_173054.2 | 10500 | Missense Mutation | CAT,CGT | H,R 3445 | NP_774959.1 |