Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGTTATCTGTCAGATGAAGGGCA[C/T]TATTGGGTGGGCCTGGATATCAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615733 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC30 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJC30 - DnaJ heat shock protein family (Hsp40) member C30 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WBSCR22 - Williams-Beuren syndrome chromosome region 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202560.2 | 286 | Silent Mutation | CAC,CAT | H,H 76 | NP_001189489.1 | |
NM_017528.4 | 286 | Silent Mutation | CAC,CAT | H,H 76 | NP_059998.2 | |
XM_006715847.1 | 286 | Silent Mutation | CAC,CAT | H,H 99 | XP_006715910.1 | |
XM_011515778.1 | 286 | Silent Mutation | CAC,CAT | H,H 99 | XP_011514080.1 | |
XM_011515779.1 | 286 | Silent Mutation | CAC,CAT | H,H 76 | XP_011514081.1 |