Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGACCATTTAGCTGGCAGTTGTC[A/G]ATATGCTTGTGGCCCACAGATCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 131170 MIM: 194624 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERV3-1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERV3-1 - endogenous retrovirus group 3 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007253.3 | 1873 | Nonsense Mutation | CGA,TGA | R,* 427 | NP_001007254.2 |
ZNF117 - zinc finger protein 117 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015852.3 | 1873 | Intron | NP_056936.2 |