Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCGCAAGGACCGGGACTCGAAG[A/G]CCCACCGCAGCCGGACTAAGTCGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603309 | ||||||||||||||||||||
Literature Links: |
CDK13 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDK13 - cyclin dependent kinase 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003718.4 | 1973 | Missense Mutation | ACC,GCC | T,A 278 | NP_003709.3 | |
NM_031267.3 | 1973 | Missense Mutation | ACC,GCC | T,A 278 | NP_112557.2 | |
XM_011515597.2 | 1973 | Missense Mutation | ACC,GCC | T,A 278 | XP_011513899.1 | |
XM_017012750.1 | 1973 | Missense Mutation | ACC,GCC | T,A 278 | XP_016868239.1 | |
XM_017012751.1 | 1973 | Missense Mutation | ACC,GCC | T,A 278 | XP_016868240.1 |