Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCAGTCTGAGCTAGCCAGCTTC[C/T]TCATGGACCACGTCTTCATCCAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608489 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GATS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GATS - GATS, stromal antigen 3 opposite strand | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178831.6 | 1010 | UTR 3 | NP_849153.3 | |||
XM_017012153.1 | 1010 | Intron | XP_016867642.1 | |||
XM_017012154.1 | 1010 | UTR 3 | XP_016867643.1 | |||
XM_017012155.1 | 1010 | UTR 3 | XP_016867644.1 | |||
XM_017012156.1 | 1010 | Intron | XP_016867645.1 | |||
XM_017012157.1 | 1010 | UTR 3 | XP_016867646.1 | |||
XM_017012158.1 | 1010 | Intron | XP_016867647.1 | |||
XM_017012159.1 | 1010 | Intron | XP_016867648.1 |
STAG3 - stromal antigen 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282716.1 | 1010 | Missense Mutation | CTC,TTC | L,F 841 | NP_001269645.1 | |
NM_001282717.1 | 1010 | Missense Mutation | CTC,TTC | L,F 841 | NP_001269646.1 | |
NM_001282718.1 | 1010 | Missense Mutation | CTC,TTC | L,F 783 | NP_001269647.1 | |
NM_012447.3 | 1010 | Missense Mutation | CTC,TTC | L,F 841 | NP_036579.2 | |
XM_011515742.1 | 1010 | Missense Mutation | CTC,TTC | L,F 841 | XP_011514044.1 | |
XM_017011683.1 | 1010 | Missense Mutation | CTC,TTC | L,F 841 | XP_016867172.1 | |
XM_017011684.1 | 1010 | Missense Mutation | CTC,TTC | L,F 841 | XP_016867173.1 | |
XM_017011685.1 | 1010 | Missense Mutation | CTC,TTC | L,F 841 | XP_016867174.1 | |
XM_017011686.1 | 1010 | Missense Mutation | CTC,TTC | L,F 841 | XP_016867175.1 | |
XM_017011687.1 | 1010 | Missense Mutation | CTC,TTC | L,F 783 | XP_016867176.1 |