Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCACCGTGAAGACGTAGCAGAA[A/G]TCCTCGGTGCTGGAGCTGCGGCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613653 MIM: 611491 | ||||||||||||||||||||
Literature Links: |
AP5Z1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AP5Z1 - adaptor related protein complex 5 zeta 1 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RADIL - Rap associating with DIL domain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018059.4 | 3083 | Silent Mutation | GAC,GAT | D,D 970 | NP_060529.4 |