Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGAAAAGCAACGTTCTTGGATG[C/T]CTGGGAGGTAGGTTCCCAGCTTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604707 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKR1B10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKR1B10 - aldo-keto reductase family 1 member B10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020299.4 | 742 | Missense Mutation | GCC,GTC | A,V 141 | NP_064695.3 | |
XM_011516416.1 | 742 | Missense Mutation | GCC,GTC | A,V 102 | XP_011514718.1 | |
XM_011516417.2 | 742 | Missense Mutation | GCC,GTC | A,V 141 | XP_011514719.1 |