Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGGCTGACCGCGATGCCAGGAC[A/G]CGCTCCCCTCCGCACCGTCCCGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606570 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPDR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPDR1 - ependymin related 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242946.1 | 390 | Missense Mutation | CAC,CGC | H,R 4 | NP_001229875.2 | |
NM_001242948.1 | 390 | Intron | NP_001229877.1 | |||
NM_017549.4 | 390 | Missense Mutation | CAC,CGC | H,R 4 | NP_060019.2 |
SFRP4 - secreted frizzled related protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |