Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGATGTTTCAAAACTGACTGAAAC[A/C]TTTGTTCATAGTAGTCAGCAACATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C7orf62 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C7orf62 - chromosome 7 open reading frame 62 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152706.3 | 397 | Missense Mutation | NP_689919.1 | |||
XM_011515904.2 | 397 | Missense Mutation | XP_011514206.1 |
ZNF804B - zinc finger protein 804B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181646.3 | 397 | Intron | NP_857597.1 |