Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGCAGCTGGCGGAAGAAGAGGAC[A/G]AGGAGCGTTTCCAGGGGTGGAAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608306 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SP8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SP8 - Sp8 transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182700.5 | 351 | Missense Mutation | TCG,TTG | S,L 55 | NP_874359.2 | |
NM_198956.3 | 351 | Missense Mutation | TCG,TTG | S,L 37 | NP_945194.1 |