Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGTGATGCGGGAGGACCTGTTC[A/G]TCGTCAGCAAGGTGCACATGGCGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616336 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKR1B15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKR1B15 - aldo-keto reductase family 1 member B15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080538.2 | 359 | Missense Mutation | ATC,GTC | I,V 103 | NP_001074007.2 | |
XM_011516241.2 | 359 | Missense Mutation | ATC,GTC | I,V 75 | XP_011514543.2 | |
XM_017012224.1 | 359 | Missense Mutation | ATC,GTC | I,V 103 | XP_016867713.1 |