Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGAATCCCATCTCCCTGCTCTGC[A/G]GCGCTATCATCATCTGGCGCTTTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615203 | ||||||||||||||||||||
Literature Links: |
RHBDD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RHBDD2 - rhomboid domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040456.1 | 278 | Missense Mutation | AGC,GGC | S,G 79 | NP_001035546.1 | |
NM_001040457.1 | 278 | UTR 5 | NP_001035547.1 | |||
XM_005250511.4 | 278 | UTR 5 | XP_005250568.1 |