Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAAGAACCACGTTGGGCCTGCTT[G/T]GTGGCTGCCACCTTGCCCCGGGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138079 MIM: 613993 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GCK - glucokinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYL7 - myosin light chain 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021223.2 | 98 | Silent Mutation | ACA,ACC | T,T 15 | NP_067046.1 | |
XM_005249817.4 | 98 | Silent Mutation | ACA,ACC | T,T 15 | XP_005249874.1 | |
XM_011515463.2 | 98 | Silent Mutation | ACA,ACC | T,T 15 | XP_011513765.1 | |
XM_011515464.2 | 98 | Silent Mutation | ACA,ACC | T,T 15 | XP_011513766.1 | |
XM_011515465.2 | 98 | Silent Mutation | ACA,ACC | T,T 15 | XP_011513767.1 | |
XM_017012478.1 | 98 | Silent Mutation | ACA,ACC | T,T 15 | XP_016867967.1 | |
XM_017012479.1 | 98 | UTR 5 | XP_016867968.1 |