Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGCCACCCAAGGCCACCAAGAA[A/G]CCCCCGTCAGGGAAGAGGCCCCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602981 MIM: 600815 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AEBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AEBP1 - AE binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129.4 | 794 | Silent Mutation | AAA,AAG | K,K 163 | NP_001120.3 | |
XM_011515162.1 | 794 | Silent Mutation | AAA,AAG | K,K 163 | XP_011513464.1 |
MIR4649 - microRNA 4649 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLD2 - DAN polymerase delta 2, accessory subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |