Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGTCCTGCAGAACTCACCTGCTC[A/G]GACGATAATTGCTGCTTCTCCTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612074 | ||||||||||||||||||||
Literature Links: |
MGC27345 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MGC27345 - uncharacterized protein MGC27345 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBM28 - RNA binding motif protein 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166135.1 | 2029 | Silent Mutation | TCC,TCT | S,S 572 | NP_001159607.1 | |
NM_018077.2 | 2029 | Silent Mutation | TCC,TCT | S,S 713 | NP_060547.2 | |
XM_011516370.2 | 2029 | Silent Mutation | TCC,TCT | S,S 747 | XP_011514672.1 | |
XM_017012389.1 | 2029 | Silent Mutation | TCC,TCT | S,S 672 | XP_016867878.1 | |
XM_017012390.1 | 2029 | Silent Mutation | TCC,TCT | S,S 638 | XP_016867879.1 |