Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTGGCGTGTTGACAATTGCCAA[C/T]GTGGATCCTTTTCCTTTGCAAAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604987 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLEC5A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLEC5A - C-type lectin domain family 5 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301167.1 | 399 | Silent Mutation | ACA,ACG | T,T 82 | NP_001288096.1 | |
NM_013252.2 | 399 | Silent Mutation | ACA,ACG | T,T 105 | NP_037384.1 | |
XM_011515995.2 | 399 | Silent Mutation | ACA,ACG | T,T 62 | XP_011514297.1 | |
XM_017011915.1 | 399 | Silent Mutation | ACA,ACG | T,T 85 | XP_016867404.1 | |
XM_017011916.1 | 399 | Silent Mutation | ACA,ACG | T,T 105 | XP_016867405.1 | |
XM_017011917.1 | 399 | Silent Mutation | ACA,ACG | T,T 82 | XP_016867406.1 |