Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGTCCTAGGAGGATTTTTCTAC[A/G]TCCGCGACTACACCCTCCTCGTCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610334 MIM: 610385 | ||||||||||||||||||||
Literature Links: |
TMEM176A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM176A - transmembrane protein 176A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018487.2 | 442 | Missense Mutation | ATC,GTC | I,V 77 | NP_060957.2 | |
XM_011516376.2 | 442 | Missense Mutation | ATC,GTC | I,V 94 | XP_011514678.1 | |
XM_011516377.2 | 442 | Missense Mutation | ATC,GTC | I,V 94 | XP_011514679.1 | |
XM_011516378.2 | 442 | Missense Mutation | ATC,GTC | I,V 94 | XP_011514680.1 | |
XM_017012393.1 | 442 | Missense Mutation | ATC,GTC | I,V 77 | XP_016867882.1 |
TMEM176B - transmembrane protein 176B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101311.1 | 442 | Intron | NP_001094781.1 | |||
NM_001101312.1 | 442 | Intron | NP_001094782.1 | |||
NM_001101314.1 | 442 | Intron | NP_001094784.1 | |||
NM_014020.3 | 442 | Intron | NP_054739.3 | |||
XM_006715933.3 | 442 | Intron | XP_006715996.2 |