Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCGGAGACCACGCTGTCCAAGGCC[C/T]GCATCAGGATGTCTGGCATGTGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614016 MIM: 602048 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DAGLB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DAGLB - diacylglycerol lipase beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142936.1 | 2107 | Missense Mutation | CAG,CGG | Q,R 517 | NP_001136408.1 | |
NM_139179.3 | 2107 | Missense Mutation | CAG,CGG | Q,R 646 | NP_631918.3 |
RAC1 - ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |