Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATCCAAGAGAAGGCTGTGATGCG[A/G]GAGGACCTGTTCATCGTCAGCAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616336 | ||||||||||||||||||||
Literature Links: |
AKR1B15 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AKR1B15 - aldo-keto reductase family 1 member B15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080538.2 | 346 | Silent Mutation | CGA,CGG | R,R 98 | NP_001074007.2 | |
XM_011516241.2 | 346 | Silent Mutation | CGA,CGG | R,R 70 | XP_011514543.2 | |
XM_017012224.1 | 346 | Silent Mutation | CGA,CGG | R,R 98 | XP_016867713.1 |