Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTTCTTTGGTGCGCAGAAGCCC[A/C]TACAAGTAGCTTCATCTGGAAACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600033 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105375401 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105375401 - uncharacterized LOC105375401 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TFPI2 - tissue factor pathway inhibitor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271003.1 | 750 | Missense Mutation | AGG,ATG | R,M 135 | NP_001257932.1 | |
NM_001271004.1 | 750 | Missense Mutation | AGG,ATG | R,M 146 | NP_001257933.1 | |
NM_006528.3 | 750 | Missense Mutation | AGG,ATG | R,M 146 | NP_006519.1 |